Assay Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026609.2 | NP_080885.1 | 2 - 3 | 128 | 84 | |
| GenBank mRNA | AK003329.1 | - | 2 - 3 | 139 | 84 | |
| AK013870.1 | - | 2 - 3 | 198 | 84 | ||
| AK075740.1 | - | 2 - 3 | 183 | 84 | ||
| AK150289.1 | - | 2 - 3 | 187 | 84 | ||
| AK153277.1 | - | 2 - 3 | 173 | 84 | ||
| AK154712.1 | - | 2 - 3 | 193 | 84 | ||
| AK159401.1 | - | 2 - 3 | 116 | 84 | ||
| AK203783.1 | - | 1 - 2 | 85 | 85 | ||
| BC004677.1 | - | 2 - 3 | 128 | 3500223 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Leprotl1
Genomic Map