Assay Details
| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029752.2 | NP_084028.1 | 1 - 2 | 331 | 90 | |
| GenBank mRNA | AK010810.1 | - | 1 - 2 | 274 | 90 | |
| AK043288.1 | - | 1 - 2 | 273 | 90 | ||
| AK044565.1 | - | 1 - 2 | 219 | 90 | ||
| AK143538.1 | - | 1 - 2 | 290 | 90 | ||
| BC039941.1 | - | 1 - 2 | 289 | 4011957 | 90 | |
More Information
Set Membership: |
|

Bri3bp
Genomic Map