Assay Details
| Gene Symbol: Scarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205082.1 | NP_001192011.1 | 2 - 3 | 497 | 87 | |
| NM_001205083.1 | NP_001192012.1 | 2 - 3 | 497 | 87 | ||
| NM_016741.2 | NP_058021.1 | 2 - 3 | 497 | 87 | ||
| GenBank mRNA | AK010622.1 | - | 2 - 3 | 422 | 87 | |
| AK028191.1 | - | 2 - 3 | 478 | 87 | ||
| AK033114.1 | - | 2 - 3 | 422 | 87 | ||
| AK145588.1 | - | 2 - 3 | 423 | 87 | ||
| AK154933.1 | - | 2 - 3 | 489 | 87 | ||
| AK158122.1 | - | 2 - 3 | 422 | 87 | ||
| AK169906.1 | - | 2 - 3 | 473 | 87 | ||
| AK171602.1 | - | 2 - 3 | 476 | 87 | ||
| BC004656.1 | - | 2 - 3 | 477 | 3498018 | 87 | |
| CT010222.1 | - | 2 - 3 | 285 | 87 | ||
| U37799.1 | - | 2 - 3 | 335 | 87 | ||
More Information
Set Membership: |
|

Scarb1
Genomic Map