Assay Details
| Gene Symbol: 4930467D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045981.1 | 2 - 3 | 222 | 125 | ||
| GenBank mRNA | AK015423.1 | - | 223 | 125 | ||
| AK015518.1 | - | 2 - 3 | 199 | 125 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
N(alpha)-acetyltransferase 11, NatA catalytic subunit |
Naa11 |
Ard1b, Ard2, C80008 |
-3.0kb | Anti-sense | Chr. 5 - 97382209 - 97392330 on NCBI Build GRCm38 |
| Downstream | glycerol kinase 2 | Gk2 |
GKTA, Gk-rs2 |
21.2kb | Anti-sense | Chr. 5 - 97455161 - 97457008 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930467D21Rik
Genomic Map