Assay Details
| Gene Symbol: A630023P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102290.1 | 2 - 3 | 270 | 76 | ||
| GenBank mRNA | AI451630.1 | - | 1 - 2 | 76 | ||
| AK041600.1 | - | 3 - 4 | 845 | 76 | ||
| AK088579.1 | - | 2 - 3 | 271 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P123655 | AI645552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P123655 | AK041600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P123655 | AK088579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P123655 | AI645552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P123655 | AK041600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P123655 | AK088579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P123655 | AI645552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P123655 | AK041600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P123655 | AK088579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P123655 | AI645552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P123655 | AK041600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P123655 | AK088579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123655 | AI645552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123655 | AK041600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123655 | AK088579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123655 | AI645552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123655 | AK041600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123655 | AK088579.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04829806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04642390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04463670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04187986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04908254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04329355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04270757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04116826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04701725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04772939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04701745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04477111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04614162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04658599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04646245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04614161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04187972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04658600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04973360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04908263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04646246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04701719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980275 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1455370_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455370_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455370_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500009832.mm.1 | TC0500001144.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009832.mm.1 | NR_102290.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009833.mm.1 | TC0500001144.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009833.mm.1 | NR_102290.1 | Refseq |
| Mouse Expression Set 430 | 1455370_at | - | - |
| Mouse Expression Set 430 | 1455370_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455370_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455370_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 630969 | - | - |

A630023P12Rik
Genomic Map