Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500028440.mm.1 |
TC0500003193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028440.mm.1 |
NM_009656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028443.mm.1 |
TC0500003193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028443.mm.1 |
NM_009656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028445.mm.1 |
TC0500003193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028445.mm.1 |
NM_009656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028439.mm.1 |
TC0500003193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028439.mm.1 |
NM_009656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028446.mm.1 |
TC0500003193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028446.mm.1 |
NM_009656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028441.mm.1 |
TC0500003193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028441.mm.1 |
NM_009656.4 |
Refseq |