Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500028334.mm.1 |
TC0500003184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028334.mm.1 |
NM_001109992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028334.mm.1 |
NM_001109992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028334.mm.1 |
NM_011202.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028334.mm.1 |
NM_011202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028335.mm.1 |
TC0500003184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028335.mm.1 |
NM_001109992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028335.mm.1 |
NM_001109992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028335.mm.1 |
NM_011202.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028335.mm.1 |
NM_011202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028336.mm.1 |
TC0500003184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028336.mm.1 |
NM_001109992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028336.mm.1 |
NM_001109992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028336.mm.1 |
NM_011202.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028336.mm.1 |
NM_011202.3 |
Refseq |