Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500012023.mm.1 |
TC0500001369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012023.mm.1 |
NM_001289543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012023.mm.1 |
NM_028227.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012025.mm.1 |
TC0500001369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012025.mm.1 |
NM_001289543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012025.mm.1 |
NM_001289544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012025.mm.1 |
NM_028227.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012027.mm.1 |
TC0500001369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012027.mm.1 |
NM_001289543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012027.mm.1 |
NM_001289544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012027.mm.1 |
NM_028227.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012026.mm.1 |
TC0500001369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012026.mm.1 |
NM_001289543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012026.mm.1 |
NM_001289544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012026.mm.1 |
NM_028227.3 |
Refseq |