Assay Details
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289543.1 | NP_001276472.1 | 7 - 8 | 1117 | 64 | |
NM_001289544.1 | NP_001276473.1 | 8 - 9 | 1441 | 64 | ||
NM_028227.3 | NP_082503.2 | 8 - 9 | 1313 | 64 | ||
GenBank mRNA | AF321920.1 | - | 8 - 9 | 1205 | 64 | |
AF321921.1 | - | 8 - 9 | 1442 | 64 | ||
AK013885.1 | - | 6 - 7 | 806 | 64 | ||
AK034212.1 | - | 3 - 4 | 246 | 64 | ||
AK143718.1 | - | 7 - 8 | 1107 | 64 | ||
BC038490.1 | - | 5 - 6 | 682 | 3599718 | 64 | |
BC062932.1 | - | 8 - 9 | 1289 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
