Assay Details
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289543.1 | NP_001276472.1 | 8 - 9 | 1226 | 71 | |
NM_001289544.1 | NP_001276473.1 | 9 - 10 | 1550 | 71 | ||
NM_028227.3 | NP_082503.2 | 9 - 10 | 1422 | 71 | ||
GenBank mRNA | AF321920.1 | - | 9 - 10 | 1314 | 71 | |
AF321921.1 | - | 9 - 10 | 1551 | 71 | ||
AK013885.1 | - | 7 - 8 | 915 | 71 | ||
AK034212.1 | - | 4 - 5 | 355 | 71 | ||
BC038490.1 | - | 6 - 7 | 791 | 3599718 | 71 | |
BC062932.1 | - | 9 - 10 | 1398 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
