Assay Details
| Gene Symbol: Loxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164311.1 | NP_001157783.1 | 5 - 6 | 808 | 108 | |
| NM_053083.3 | NP_444313.3 | 5 - 6 | 808 | 108 | ||
| GenBank mRNA | AF338440.1 | - | 4 - 5 | 703 | 108 | |
| AK014331.1 | - | 5 - 6 | 808 | 108 | ||
| BC068307.1 | - | 5 - 6 | 750 | 30059745 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Loxl4
Genomic Map