Assay Details
| Gene Symbol: Loxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164311.1 | NP_001157783.1 | 6 - 7 | 1029 | 68 | |
| GenBank mRNA | AK014331.1 | - | 6 - 7 | 1029 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Loxl4
Genomic Map