Assay Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019734.2 | NP_062708.1 | 11 - 12 | 1023 | 69 | |
| GenBank mRNA | AF157500.1 | - | 11 - 12 | 953 | 69 | |
| AF352179.1 | - | 11 - 12 | 965 | 69 | ||
| AK075666.1 | - | 11 - 12 | 938 | 69 | ||
| AK075919.1 | - | 11 - 12 | 977 | 69 | ||
| AK077721.1 | - | 11 - 12 | 976 | 69 | ||
| AK090314.1 | - | 11 - 12 | 973 | 69 | ||
| AK150567.1 | - | 11 - 12 | 935 | 69 | ||
| AK151208.1 | - | 11 - 12 | 1024 | 69 | ||
| AK151312.1 | - | 11 - 12 | 935 | 69 | ||
| AK151478.1 | - | 11 - 12 | 1014 | 69 | ||
| AK152305.1 | - | 11 - 12 | 975 | 69 | ||
| AK152811.1 | - | 11 - 12 | 936 | 69 | ||
| AK152920.1 | - | 11 - 12 | 983 | 69 | ||
| AK153101.1 | - | 11 - 12 | 983 | 69 | ||
| AK159556.1 | - | 11 - 12 | 945 | 69 | ||
| AK166757.1 | - | 11 - 12 | 955 | 69 | ||
| BC003204.1 | - | 11 - 12 | 943 | 3587042 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah1
Genomic Map