Assay Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019734.2 | NP_062708.1 | 5 - 6 | 487 | 156 | |
| GenBank mRNA | AF157500.1 | - | 5 - 6 | 417 | 156 | |
| AF352179.1 | - | 5 - 6 | 429 | 156 | ||
| AK075666.1 | - | 5 - 6 | 402 | 156 | ||
| AK075919.1 | - | 5 - 6 | 441 | 156 | ||
| AK077721.1 | - | 5 - 6 | 440 | 156 | ||
| AK090314.1 | - | 5 - 6 | 437 | 156 | ||
| AK150567.1 | - | 5 - 6 | 400 | 156 | ||
| AK151208.1 | - | 5 - 6 | 488 | 156 | ||
| AK151312.1 | - | 5 - 6 | 400 | 156 | ||
| AK151478.1 | - | 5 - 6 | 478 | 156 | ||
| AK152305.1 | - | 5 - 6 | 439 | 156 | ||
| AK152811.1 | - | 5 - 6 | 400 | 156 | ||
| AK152920.1 | - | 5 - 6 | 447 | 156 | ||
| AK153101.1 | - | 5 - 6 | 447 | 156 | ||
| AK153297.1 | - | 5 - 6 | 401 | 156 | ||
| AK159556.1 | - | 5 - 6 | 409 | 156 | ||
| AK166757.1 | - | 5 - 6 | 419 | 156 | ||
| BC003204.1 | - | 5 - 6 | 407 | 3587042 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah1
Genomic Map