Assay Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019734.2 | NP_062708.1 | 7 - 8 | 611 | 76 | |
| GenBank mRNA | AF157500.1 | - | 7 - 8 | 541 | 76 | |
| AF352179.1 | - | 7 - 8 | 553 | 76 | ||
| AK075666.1 | - | 7 - 8 | 526 | 76 | ||
| AK075919.1 | - | 7 - 8 | 565 | 76 | ||
| AK077721.1 | - | 7 - 8 | 564 | 76 | ||
| AK090314.1 | - | 7 - 8 | 561 | 76 | ||
| AK150567.1 | - | 7 - 8 | 524 | 76 | ||
| AK151208.1 | - | 7 - 8 | 612 | 76 | ||
| AK151312.1 | - | 7 - 8 | 524 | 76 | ||
| AK151478.1 | - | 7 - 8 | 602 | 76 | ||
| AK152305.1 | - | 7 - 8 | 563 | 76 | ||
| AK152811.1 | - | 7 - 8 | 76 | |||
| AK152920.1 | - | 7 - 8 | 571 | 76 | ||
| AK153101.1 | - | 7 - 8 | 571 | 76 | ||
| AK153297.1 | - | 7 - 8 | 525 | 76 | ||
| AK159556.1 | - | 7 - 8 | 533 | 76 | ||
| AK166757.1 | - | 7 - 8 | 543 | 76 | ||
| BC003204.1 | - | 7 - 8 | 531 | 3587042 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah1
Genomic Map