Assay Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019734.2 | NP_062708.1 | 9 - 10 | 811 | 115 | |
| GenBank mRNA | AF157500.1 | - | 9 - 10 | 741 | 115 | |
| AF352179.1 | - | 9 - 10 | 753 | 115 | ||
| AK075666.1 | - | 9 - 10 | 726 | 115 | ||
| AK075919.1 | - | 9 - 10 | 765 | 115 | ||
| AK077721.1 | - | 9 - 10 | 764 | 115 | ||
| AK090314.1 | - | 9 - 10 | 761 | 115 | ||
| AK150567.1 | - | 9 - 10 | 723 | 115 | ||
| AK151208.1 | - | 9 - 10 | 812 | 115 | ||
| AK151312.1 | - | 9 - 10 | 723 | 115 | ||
| AK151478.1 | - | 9 - 10 | 802 | 115 | ||
| AK152305.1 | - | 9 - 10 | 763 | 115 | ||
| AK152811.1 | - | 9 - 10 | 723 | 115 | ||
| AK152920.1 | - | 9 - 10 | 771 | 115 | ||
| AK153101.1 | - | 9 - 10 | 771 | 115 | ||
| AK153297.1 | - | 9 - 10 | 725 | 115 | ||
| AK159556.1 | - | 9 - 10 | 733 | 115 | ||
| AK166757.1 | - | 9 - 10 | 743 | 115 | ||
| BC003204.1 | - | 9 - 10 | 731 | 3587042 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah1
Genomic Map