Assay Details
| Gene Symbol: Dgkq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199011.1 | NP_950176.1 | 20 - 21 | 2513 | 108 | |
| GenBank mRNA | AK043933.1 | - | 2680 | 108 | ||
| AK048764.1 | - | 19 - 20 | 2831 | 108 | ||
| AK132151.1 | - | 19 - 20 | 2420 | 108 | ||
| AK134745.1 | - | 7 - 8 | 1576 | 108 | ||
| AK170840.1 | - | 17 - 18 | 2479 | 108 | ||
| BC062929.1 | - | 20 - 21 | 2513 | 6855058 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgkq
Genomic Map