Assay Details
Gene Symbol: Mfsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172883.2 | NP_766471.1 | 4 - 5 | 621 | 65 | |
GenBank mRNA | AK029021.1 | - | 4 - 5 | 621 | 65 | |
AK087226.1 | - | 4 - 5 | 831 | 65 | ||
AK171069.1 | - | 3 - 4 | 3253 | 65 | ||
BC094512.1 | - | 2 - 3 | 233 | 2616131 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC094512.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
