Assay Details
| Gene Symbol: Mtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB221622.1 | - | 10 - 11 | 1005 | 85 | |
| BC050826.1 | - | 13 - 14 | 1557 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Mtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB221622.1 | - | 10 - 11 | 1005 | 85 | |
| BC050826.1 | - | 13 - 14 | 1557 | 85 | ||
Set Membership: |
|