Assay Details
| Gene Symbol: Pigg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081234.1 | NP_001074703.1 | 9 - 10 | 2042 | 66 | |
| GenBank mRNA | BC051059.1 | - | 3 - 4 | 923 | 66 | |
More Information
Set Membership: |
|

Pigg
Genomic Map