Assay Details
| Gene Symbol: Pigg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081234.1 | NP_001074703.1 | 11 - 12 | 2548 | 65 | |
| GenBank mRNA | BC051059.1 | - | 5 - 6 | 1429 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigg
Genomic Map