Assay Details
Gene Symbol: Pigg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081234.1 | NP_001074703.1 | 5 - 6 | 894 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
