Assay Details
Gene Symbol: Fra10ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081075.1 | NP_001074544.1 | 3 - 4 | 343 | 103 | |
GenBank mRNA | AK017666.1 | - | 3 - 4 | 405 | 103 | |
AK077572.1 | - | 3 - 4 | 316 | 103 | ||
AK172241.1 | - | 3 - 4 | 350 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
