Assay Details
| Gene Symbol: Cwf19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081077.1 | NP_001074546.1 | 12 - 13 | 1461 | 99 | |
| GenBank mRNA | AK087867.1 | - | 6 - 7 | 734 | 99 | |
| AK088543.1 | - | 14 - 15 | 2703 | 99 | ||
| AK139654.1 | - | 12 - 13 | 1462 | 99 | ||
| BC037640.1 | - | 12 - 13 | 1439 | 5096937 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cwf19l1
Genomic Map