Assay Details
| Gene Symbol: Cwf19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081077.1 | NP_001074546.1 | 5 - 6 | 587 | 79 | |
| GenBank mRNA | AK012163.1 | - | 5 - 6 | 571 | 79 | |
| AK088543.1 | - | 7 - 8 | 1829 | 79 | ||
| AK139654.1 | - | 5 - 6 | 588 | 79 | ||
| BC027553.1 | - | 5 - 6 | 572 | 79 | ||
| BC037640.1 | - | 5 - 6 | 565 | 5096937 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cwf19l1
Genomic Map