Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300021571.mm.1 |
NM_001204979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021571.mm.1 |
NM_001204979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021571.mm.1 |
NM_011319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021571.mm.1 |
NM_011319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021571.mm.1 |
TC0300002742.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021573.mm.1 |
NM_001204979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021573.mm.1 |
NM_001204979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021573.mm.1 |
NM_011319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021573.mm.1 |
NM_011319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021573.mm.1 |
TC0300002742.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021568.mm.1 |
NM_001204979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021568.mm.1 |
NM_001204979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021568.mm.1 |
NM_011319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021568.mm.1 |
NM_011319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021568.mm.1 |
TC0300002742.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021567.mm.1 |
NM_001204979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021567.mm.1 |
NM_001204979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021567.mm.1 |
NM_011319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021567.mm.1 |
NM_011319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021567.mm.1 |
TC0300002742.mm.1 |
MTA TC |