Assay Details
Gene Symbol: Denr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026603.4 | NP_080879.1 | 4 - 5 | 307 | 110 | |
GenBank mRNA | AK017647.1 | - | 4 - 5 | 336 | 110 | |
AK018762.1 | - | 4 - 5 | 308 | 110 | ||
AK135127.1 | - | 4 - 5 | 343 | 110 | ||
BC025143.1 | - | 4 - 5 | 291 | 110 | ||
BC043922.1 | - | 4 - 5 | 275 | 5373203 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
