Assay Details
Gene Symbol: Abcb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019875.2 | NP_063928.2 | 5 - 6 | 1281 | 99 | |
GenBank mRNA | AB045382.2 | - | 4 - 5 | 1101 | 99 | |
AB093298.1 | - | 5 - 6 | 1263 | 99 | ||
AF216495.1 | - | 5 - 6 | 1274 | 99 | ||
AK044140.1 | - | 5 - 6 | 1259 | 99 | ||
BC053014.1 | - | 5 - 6 | 1234 | 5686698 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
