Assay Details
| Gene Symbol: Gtf2h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181410.3 | NP_852075.1 | 3 - 4 | 245 | 56 | |
| GenBank mRNA | AK050227.1 | - | 3 - 4 | 229 | 56 | |
| AK133941.1 | - | 3 - 4 | 216 | 56 | ||
| BC017515.1 | - | 3 - 4 | 222 | 4504077 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2h3
Genomic Map