Assay Details
| Gene Symbol: Gtf2h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181410.3 | NP_852075.1 | 8 - 9 | 607 | 75 | |
| GenBank mRNA | AK017176.1 | - | 1 - 2 | 570 | 75 | |
| AK050227.1 | - | 8 - 9 | 591 | 75 | ||
| AK133941.1 | - | 8 - 9 | 578 | 75 | ||
| BC017515.1 | - | 8 - 9 | 584 | 4504077 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2h3
Genomic Map