Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500012771.mm.1 |
TC0500003713.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012771.mm.1 |
NM_001134767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012771.mm.1 |
NM_001134767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012773.mm.1 |
TC0500003713.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012773.mm.1 |
NM_001134767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012773.mm.1 |
NM_001134767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012775.mm.1 |
TC0500003713.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012775.mm.1 |
NM_001134767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012775.mm.1 |
NM_001134767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012772.mm.1 |
TC0500003713.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012772.mm.1 |
NM_001134767.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012772.mm.1 |
NM_001134767.1 |
Refseq |