Assay Details
| Gene Symbol: Arl6ip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144509.2 | NP_653092.1 | 4 - 5 | 973 | 79 | |
| GenBank mRNA | AB035383.1 | - | 4 - 5 | 973 | 79 | |
| AK133934.1 | - | 4 - 5 | 789 | 79 | ||
| BC063748.1 | - | 4 - 5 | 940 | 6819350 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059369 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059367 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059365 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059364 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059366 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P454666 | AF133911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P454666 | AF133911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137157 | BC063748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137157 | NM_144509.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137157 | AB035383.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889228 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661418 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813863 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125363 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121311 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133938 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109324 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155536 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P454666 | AF133911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P454666 | AF133911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137157 | BC063748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137157 | NM_144509.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137157 | AB035383.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059364 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05729356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109324 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01210559 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05729491 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133938 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05729417 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155536 | - | - |
| Whole Mouse Genome G4122A | A_52_P533065 | BC063748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533065 | NM_144509.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533065 | AB035383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P533065 | AK133934.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454666 | AF133911.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454666 | AF133911.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137157 | BC063748.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137157 | NM_144509.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137157 | AB035383.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109324 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155536 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01210559 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04711915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04711907 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04711913 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04711904 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04531912 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04219058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059366 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059365 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059364 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059367 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04711915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04711904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04531901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059376 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525722 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525718 | - | - |
| Mouse Genome 430 2.0 Array | 1424285_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424285_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65105_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65105_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500012889.mm.1 | TC0500001437.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012889.mm.1 | NM_144509.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012889.mm.1 | NM_144509.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012888.mm.1 | TC0500001437.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012888.mm.1 | NM_144509.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012888.mm.1 | NM_144509.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012890.mm.1 | TC0500001437.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012890.mm.1 | NM_144509.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012890.mm.1 | NM_144509.2 | Refseq |
| Mouse Expression Set 430 | 1424285_PM_s_at | - | - |
| Mouse Expression Set 430 | 1424285_s_at | - | - |
| Murine Genome U74v2 Set | 95136_PM_at | - | - |
| Murine Genome U74v2 Set | 95136_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424285_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424285_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 813866 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5690193 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690193 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3800021 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690193 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800021 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2740176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2760116 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2760116 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2740176 | - | - |
| mouse-6 v1.1 (Illumina) | scl0004030.1_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl27224.3.7_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690193 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800021 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004030.1_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27224.3.7_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800021 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3034691 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2740176 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3108440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2760116 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3034691 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2760116 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3108440 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2740176 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0004030.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl27224.3.7_29-S | - | - |

Arl6ip4
Genomic Map