Assay Details
| Gene Symbol: Ccdc62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134767.1 | NP_001128239.1 | 9 - 10 | 1839 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999551 | NM_001134767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999551 | AK135815.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 60780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556997 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203497 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614804 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146625 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999551 | NM_001134767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999551 | AK135815.1 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525673 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525675 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525672 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525667 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525666 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525674 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525668 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525676 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458644_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525665 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500012786.mm.1 | TC0500003713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012786.mm.1 | NM_001134767.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012786.mm.1 | NM_001134767.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012787.mm.1 | TC0500003713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012787.mm.1 | NM_001134767.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012787.mm.1 | NM_001134767.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012790.mm.1 | TC0500003713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012790.mm.1 | NM_001134767.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012790.mm.1 | NM_001134767.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012789.mm.1 | TC0500003713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012789.mm.1 | NM_001134767.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012789.mm.1 | NM_001134767.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012793.mm.1 | TC0500003713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012793.mm.1 | NM_001134767.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012793.mm.1 | NM_001134767.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012792.mm.1 | TC0500003713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012792.mm.1 | NM_001134767.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012792.mm.1 | NM_001134767.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012794.mm.1 | TC0500003713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012794.mm.1 | NM_001134767.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012794.mm.1 | NM_001134767.1 | Refseq |
| Murine Genome U74v2 Set | 135170_PM_at | - | - |
| Murine Genome U74v2 Set | 135170_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458644_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458644_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231259 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251075 | - | - |

Ccdc62
Genomic Map