Assay Details
| Gene Symbol: Ampd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289719.1 | NP_001276648.1 | 8 - 9 | 1136 | 77 | |
| NM_001289720.1 | NP_001276649.1 | 7 - 8 | 953 | 77 | ||
| NM_028779.5 | NP_083055.1 | 7 - 8 | 1222 | 77 | ||
| GenBank mRNA | AK004759.2 | - | 8 - 9 | 1121 | 77 | |
| AK034844.1 | - | 7 - 8 | 926 | 77 | ||
| AK169980.1 | - | 8 - 9 | 1122 | 77 | ||
| BC049119.1 | - | 7 - 8 | 1183 | 6812571 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ampd2
Genomic Map