Assay Details
Gene Symbol: 2310002F09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_077063.1 | 2 - 3 | 229 | 173 | ||
GenBank mRNA | AK009091.1 | - | 2 - 3 | 230 | 173 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kallikrein related-peptidase 13 | Klk13 |
Egfbp-2, mGk-13 |
-19.2kb | Anti-sense | Chr. 7 - 43712567 - 43727864 on NCBI Build GRCm38 |
Downstream | kallikrein related-peptidase 12 | Klk12 |
2310008B01Rik, AI324852, KLK-L5 |
19.9kb | Anti-sense | Chr. 7 - 43768664 - 43774704 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2292737 | AK086402.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160588 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK006129.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK009463.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK009475.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK009542.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK010001.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK012062.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK012259.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | AK141494.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923291 | NM_001004190.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197724 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357515 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450798 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615853 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 70600 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108577 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694758 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491281 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859102 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769603 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571783 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22705 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803990 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637852 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572343 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338831 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364655 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578324 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437074 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756744 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843944 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | AK004805.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | AK005263.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | AK081598.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | AK159237.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | AK168500.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | AK220236.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | BC011466.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | BC098227.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | BC119133.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | BC120841.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551174 | NM_026267.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447330 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952986 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429540 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899099 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818960 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59818 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886816 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446186 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520774 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20917 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68651 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71229 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74422 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49255 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134586 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14273 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143719 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175122 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2292737 | AK086402.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178192 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178221 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143719 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178145 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178231 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178304 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178136 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178236 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178139 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175122 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01505213 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01505174 | - | - |
Whole Mouse Genome G4122A | A_52_P415836 | AK086402.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P415836 | AK009091.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103002 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06178221 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01505174 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143719 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175122 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06178145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25083866 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05902826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05902820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25083867 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25083868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25083874 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25083870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25083875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25083865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05902821 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0700026165.mm.1 | TC0700002952.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700026165.mm.1 | NR_077063.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700026164.mm.1 | TC0700002952.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700026164.mm.1 | NR_077063.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700026163.mm.1 | TC0700002952.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700026163.mm.1 | NR_077063.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 802229 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103610026 | - | - |
