Assay Details
Gene Symbol: Fam109a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175474.3 | NP_780683.1 | 1 - 2 | 96 | 79 | |
GenBank mRNA | AK033618.1 | - | 1 - 2 | 97 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK033618.1, NM_175474.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
