Assay Details
Gene Symbol: Fam109a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175474.3 | NP_780683.1 | 2 - 3 | 174 | 91 | |
GenBank mRNA | AK033618.1 | - | 2 - 3 | 175 | 91 | |
BC120681.1 | - | 1 - 2 | 74 | 40129604 | 91 | |
BC120683.1 | - | 1 - 2 | 74 | 40129606 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK033618.1, BC120681.1, BC120683.1, NM_175474.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
