Assay Details
Gene Symbol: Fam109a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175474.3 | NP_780683.1 | 3 - 4 | 283 | 75 | |
GenBank mRNA | AK033618.1 | - | 3 - 4 | 284 | 75 | |
AK039192.1 | - | 2 - 3 | 231 | 75 | ||
BC120681.1 | - | 2 - 3 | 183 | 40129604 | 75 | |
BC120683.1 | - | 2 - 3 | 183 | 40129606 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK033618.1, AK039192.1, BC120681.1, BC120683.1, NM_175474.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046892 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046894 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046888 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046860 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046862 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046861 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046893 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046864 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046889 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046890 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046863 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046891 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | BC095929.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | AK039192.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | AK081460.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | AK033618.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | NM_175474.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | BC095929.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | AK039192.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | AK081460.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | AK033618.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466829 | NM_175474.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185905 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183036 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620773 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481314 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569623 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15207 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146392 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176748 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | BC095929.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | AK039192.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | AK081460.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | AK033618.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | NM_175474.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | BC095929.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | AK039192.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | AK081460.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | AK033618.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466829 | NM_175474.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24046867 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146392 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176748 | - | - |
Whole Mouse Genome G4122A | A_51_P466829 | BC095929.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | AK039192.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | AK081460.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | AK033618.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | NM_175474.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | BC095929.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | AK039192.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | AK081460.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | AK033618.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P466829 | NM_175474.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106100 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176748 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146392 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04596043 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04596042 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04820333 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046889 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046892 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046860 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046863 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046864 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046862 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046861 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046888 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046893 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046894 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046891 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24046890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24046873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04213087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04200356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046903 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04200359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04791990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04200349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04213092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046848 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04389282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04773375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04200357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046864 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220417 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04130691 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04200351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04656906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04820333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046856 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046897 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04860757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046858 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04213107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046832 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04213116 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04392846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04130693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04837517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04130694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04773367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04200353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046841 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04475055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220415 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04200358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04220416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24046905 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10525337 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525339 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525341 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525338 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525340 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454662_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10525336 | - | - |
Mouse Genome 430 2.0 Array | 1454662_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454662_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231717_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231717_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500012123.mm.1 | TC0500001374.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500012123.mm.1 | NM_175474.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500012123.mm.1 | NM_175474.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500012125.mm.1 | TC0500001374.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500012125.mm.1 | NM_175474.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500012125.mm.1 | NM_175474.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500012124.mm.1 | TC0500001374.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500012124.mm.1 | NM_175474.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500012124.mm.1 | NM_175474.3 | Refseq |
Mouse Expression Set 430 | 1454662_PM_at | - | - |
Mouse Expression Set 430 | 1454662_at | - | - |
Murine Genome U74v2 Set | 160598_PM_at | - | - |
Murine Genome U74v2 Set | 160598_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454662_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454662_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 913333 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105700039 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 105700039 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5270435 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005270435 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2668178 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2668178 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1247128 | - | - |
mouse-6 v1.1 (Illumina) | scl00231717.2_326-S | - | - |
mouse-6 v1.1 (Illumina) | scl27269.5_229-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5270435 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105700039 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00231717.2_326-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270435 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105700039 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2668178 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2668178 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00231717.2_326-S | - | - |
