Assay Details
| Gene Symbol: Mzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290452.1 | NP_001277381.1 | 2 - 3 | 725 | 61 | |
| GenBank mRNA | BC129904.1 | - | 2 - 3 | 475 | 40111654 | 61 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC129904.1, NM_001290452.1 transcripts.
Set Membership: |
|

Mzf1
Genomic Map