Assay Details
| Gene Symbol: Dram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025582.2 | NP_001020753.1 | 4 - 5 | 459 | 109 | |
| NM_001286986.1 | NP_001273915.1 | 4 - 5 | 456 | 109 | ||
| NM_001286987.1 | NP_001273916.1 | 3 - 4 | 334 | 109 | ||
| NM_026013.3 | NP_080289.1 | 4 - 5 | 459 | 109 | ||
| GenBank mRNA | AI875295.1 | - | 4 - 5 | 459 | 109 | |
| AK008532.1 | - | 4 - 5 | 383 | 109 | ||
| AK009940.1 | - | 4 - 5 | 396 | 109 | ||
| AK012044.1 | - | 4 - 5 | 376 | 109 | ||
| AK015888.1 | - | 4 - 5 | 422 | 109 | ||
| AK052824.1 | - | 3 - 4 | 268 | 109 | ||
| AK172455.1 | - | 4 - 5 | 426 | 109 | ||
| BC024458.1 | - | 4 - 5 | 385 | 4975302 | 109 | |
| BI738450.1 | - | 4 - 5 | 427 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dram2
Genomic Map