Assay Details
| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145467.2 | NP_663442.2 | 6 - 7 | 1032 | 79 | |
| GenBank mRNA | AK029068.1 | - | 6 - 7 | 999 | 79 | |
| AK051773.1 | - | 1070 | 79 | |||
| BC020152.1 | - | 6 - 7 | 999 | 3708340 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgbl1
Genomic Map