Assay Details
| Gene Symbol: Cpsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291248.1 | NP_001278177.1 | 2 - 3 | 298 | 74 | |
| NM_001291249.1 | NP_001278178.1 | 2 - 3 | 298 | 74 | ||
| NM_178576.3 | NP_848671.1 | 2 - 3 | 298 | 74 | ||
| GenBank mRNA | AF033201.1 | - | 2 - 3 | 115 | 74 | |
| AK046064.1 | - | 2 - 3 | 288 | 74 | ||
| CN723345.1 | - | 2 - 3 | 280 | 74 | ||
| EU588393.1 | - | 2 - 3 | 154 | 74 | ||
| EU588394.1 | - | 2 - 3 | 154 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf4
Genomic Map