Assay Details
| Gene Symbol: Zkscan14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023322.2 | NP_075811.2 | 2 - 3 | 722 | 124 | |
| GenBank mRNA | AK012630.1 | - | 1 - 2 | 123 | ||
| AK013258.1 | - | 2 - 3 | 710 | 124 | ||
| AK153972.1 | - | 2 - 3 | 705 | 124 | ||
| BC141244.1 | - | 2 - 3 | 648 | 9055856 | 124 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012630.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164833 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | U62906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | U62906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438620 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33470 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172036 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | U62906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | U62906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | U62906.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | U62906.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164829 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05779276 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172036 | - | - |
| Whole Mouse Genome G4122A | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | U62906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | BC141244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | U62906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | AK153972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | AK013258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P399614 | NM_023322.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130653 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172036 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04684233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04608007 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04275872 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04275877 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04275876 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04608006 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04684237 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04275875 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04684232 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04684239 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04275864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04275868 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04472683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04859288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04158041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04105696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04472682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04684239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04173559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04158049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04722814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04474534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04981837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535656 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535657 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535658 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418607_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427948_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452610_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535653 | - | - |
| Mouse Genome 430 2.0 Array | 1427948_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427948_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1418607_at | - | - |
| Mouse Genome 430 2.0 Array | 1452610_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418607_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452610_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67235_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67235_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032961.mm.1 | TC0500003592.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032961.mm.1 | NM_023322.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032961.mm.1 | NM_023322.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032962.mm.1 | TC0500003592.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032962.mm.1 | NM_023322.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032962.mm.1 | NM_023322.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032963.mm.1 | TC0500003592.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032963.mm.1 | NM_023322.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032963.mm.1 | NM_023322.2 | Refseq |
| Mouse Expression Set 430 | 1418607_PM_at | - | - |
| Mouse Expression Set 430 | 1418607_at | - | - |
| Murine Genome U74v2 Set | 104010_at | - | - |
| Murine Genome U74v2 Set | 104010_PM_at | - | - |
| Murine Genome U74v2 Set | 167585_at | - | - |
| Murine Genome U74v2 Set | 167585_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418607_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418607_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427948_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418607_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427948_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418607_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452610_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452610_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1740577 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740577 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2497349 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2497349 | - | - |
| mouse-6 v1.1 (Illumina) | scl25783.3.1_76-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25783.3.1_76-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2497349 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2497349 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25783.3.1_76-S | - | - |

Zkscan14
Genomic Map