Assay Details
| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016798.3 | NP_058078.1 | 5 - 6 | 744 | 62 | |
| GenBank mRNA | AF002283.1 | - | 5 - 6 | 768 | 62 | |
| AK078421.1 | - | 5 - 6 | 744 | 62 | ||
| BC070418.1 | - | 5 - 6 | 710 | 5714572 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdlim3
Genomic Map