Assay Details
Gene Symbol: Ly6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164036.1 | NP_001157508.1 | 1 - 2 | 650 | 99 | |
NM_001164037.1 | NP_001157509.1 | 2 - 3 | 523 | 99 | ||
NM_001164038.1 | NP_001157510.1 | 2 - 3 | 439 | 99 | ||
GenBank mRNA | AK014207.1 | - | 1 - 2 | 105 | 99 | |
BC003926.1 | - | 1 - 2 | 53 | 3485382 | 99 | |
BC005684.1 | - | 1 - 2 | 85 | 3155116 | 99 | |
BC040691.1 | - | 1 - 2 | 70 | 5025135 | 99 | |
BC053523.1 | - | 1 - 2 | 53 | 4205673 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK014207.1,BC003926.1,BC005684.1,BC040691.1,BC053523.1,NM_001164036.1,NM_001164037.1,NM_001164038.1 transcripts.
This assay is in the 5'UTR region of the AK014207.1,BC003926.1,BC005684.1,BC040691.1,BC053523.1,NM_001164036.1,NM_001164037.1,NM_001164038.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
