Assay Details
| Gene Symbol: Khdrbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014353.1 | - | 10 - 11 | 1468 | 76 | |
| AK132792.1 | - | 9 - 10 | 2595 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdrbs3| Gene Symbol: Khdrbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014353.1 | - | 10 - 11 | 1468 | 76 | |
| AK132792.1 | - | 9 - 10 | 2595 | 76 | ||
Set Membership: |
|