Assay Details
Gene Symbol: Khdrbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010158.2 | NP_034288.2 | 3 - 4 | 654 | 66 | |
GenBank mRNA | AF079763.1 | - | 3 - 4 | 662 | 66 | |
AF099092.1 | - | 3 - 4 | 665 | 66 | ||
AK132792.1 | - | 3 - 4 | 355 | 66 | ||
BC031507.1 | - | 3 - 4 | 400 | 4500976 | 66 | |
BC057577.1 | - | 3 - 4 | 638 | 6414938 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
