Assay Details
| Gene Symbol: Khdrbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010158.2 | NP_034288.2 | 4 - 5 | 798 | 70 | |
| GenBank mRNA | AF099092.1 | - | 4 - 5 | 809 | 70 | |
| AK132792.1 | - | 4 - 5 | 499 | 70 | ||
| AK160564.1 | - | 1 - 2 | 145 | 70 | ||
| BC031507.1 | - | 4 - 5 | 544 | 4500976 | 70 | |
| BC057577.1 | - | 4 - 5 | 782 | 6414938 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Khdrbs3
Genomic Map