Assay Details
Gene Symbol: Fbxl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013909.2 | NP_038937.2 | 7 - 8 | 1225 | 59 | |
GenBank mRNA | AB041571.1 | - | 6 - 7 | 1463 | 59 | |
AF176522.1 | - | 7 - 8 | 1213 | 59 | ||
AK040458.1 | - | 5 - 6 | 1605 | 59 | ||
AK052585.1 | - | 4 - 5 | 1638 | 59 | ||
BC011485.1 | - | 5 - 6 | 1598 | 59 | ||
BC026818.1 | - | 5 - 6 | 1592 | 59 | ||
BC032263.1 | - | 2 - 3 | 59 | |||
BC044784.2 | - | 6 - 7 | 1425 | 59 | ||
BC054377.1 | - | 6 - 7 | 1046 | 6315438 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348996 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348997 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348999 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348995 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348994 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC054377.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC026818.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC032263.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC044784.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | AK052585.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC054377.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC026818.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC032263.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | BC044784.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P429246 | AK052585.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330515 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104793 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC054377.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC026818.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC032263.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC044784.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | AK052585.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC054377.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC026818.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC032263.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | BC044784.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P429246 | AK052585.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348980 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348989 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348962 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348960 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30348972 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104793 | - | - |
Whole Mouse Genome G4122A | A_51_P429246 | BC054377.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | BC026818.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | BC032263.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | BC044784.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | AK052585.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | BC054377.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | BC026818.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | BC032263.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | BC044784.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P429246 | AK052585.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100764 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104793 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12115191 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12115223 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12115226 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12115192 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12115210 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348997 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348996 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348995 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348994 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30348999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30348981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348949 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348976 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348954 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348952 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12464483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348957 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348979 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348967 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348993 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348953 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348955 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30348972 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10429958 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429963 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429964 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429967 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429961 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429966 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429962 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429960 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429965 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429959 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449849_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10429957 | - | - |
Mouse Genome 430 2.0 Array | 1449849_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1449849_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30840_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30840_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500013695.mm.1 | NM_013909.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500013695.mm.1 | NM_013909.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500013695.mm.1 | TC1500001780.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500013694.mm.1 | NM_013909.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500013694.mm.1 | NM_013909.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500013694.mm.1 | TC1500001780.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449849_PM_a_at | - | - |
Mouse Expression Set 430 | 1449849_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1449849_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1449849_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449849_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449849_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2190739 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6110537 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006110537 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002190739 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769546 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216206 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2769546 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1216206 | - | - |
mouse-6 v1.1 (Illumina) | scl47039.3.1_28-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002466.1_124-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6110537 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2190739 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl47039.3.1_28-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002466.1_124-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110537 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190739 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2935032 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1216206 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216206 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2935032 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47039.3.1_28-S | - | - |
