| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700002084.mm.1 |
NM_001164676.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002084.mm.1 |
NM_001164676.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002084.mm.1 |
TC1700000279.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002090.mm.1 |
NM_001164676.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002090.mm.1 |
NM_001164676.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002090.mm.1 |
TC1700000279.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002089.mm.1 |
NM_001164676.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002089.mm.1 |
NM_001164676.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002089.mm.1 |
TC1700000279.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002091.mm.1 |
NM_001164676.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002091.mm.1 |
NM_001164676.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002091.mm.1 |
TC1700000279.mm.1 |
MTA TC |